Learn More
Fisherbrand™ Disposable Plain-End Flint Glass Tubes with Color-Coded Label Area
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
169.32€
Specifications
Material | Flint Glass |
---|---|
Disposable | Yes |
Sterility | Non-sterile |
Closure Type | KIM-KAP |
Product Type | Disposable Culture Tube |
Product Code | Brand | Color | Diameter (Metric) | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Diameter (Metric) | Length (Metric) | Capacity (Metric) | Price | Quantity & Availability | ||
11724426
|
Fisherbrand™
|
White | 13 mm | 100 mm | 10 mL |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||
Description
Eliminate the problem of lost or peeling labels with these disposable, color-coded tubes.
- Fused-in, textured, color patches accept any type of ink or pencil
- Expose to heat, steam, hours in a water bath, without affecting label
- Marking area is large enough for all necessary information—1.37 in. (for 10 and 12mm O.D. tubes) or 2 in. (for 13 and 16mm O.D. tubes) long labeling area in five pastel colors
- Round bottom
- Flint or borosilicate glass
Specifications
Flint Glass | |
Non-sterile | |
Disposable Culture Tube |
Yes | |
KIM-KAP |