missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Yellow Indicator, Honeywell Fluka™

Catalog Number 15623910
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
60-11-7
C14H15N3
225.30
MFCD00008308
JCYPECIVGRXBMO-UHFFFAOYSA-N
6053
CHEBI:17903
N,N-dimethyl-4-phenyldiazenylaniline
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1
This item is not returnable. View return policy
60-11-7
C14H15N3
225.30
MFCD00008308
JCYPECIVGRXBMO-UHFFFAOYSA-N
6053
CHEBI:17903
N,N-dimethyl-4-phenyldiazenylaniline
CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1

Indicator

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 111°C
Linear Formula C6H5N=NC6H4N(CH3)2
UN Number UN2811
Beilstein 746016
Solubility Information Insoluble in water
Formula Weight 225.29g/mol
Odor Odourless
Packaging Glass Bottle
Color Yellow
pH 5 to 7 (at 10.00 g/L 20°C)
Quantity 100 g
Chemical Name or Material Dimethyl Yellow
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'