missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Yellow Indicator, Honeywell Fluka™
72.52 EUR valid until 2024-04-26
Use promo code "21687" to get your promotional price.
Use promo code "21687" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Melting Point | 111°C |
Linear Formula | C6H5N=NC6H4N(CH3)2 |
UN Number | UN2811 |
Beilstein | 746016 |
Solubility Information | Insoluble in water |
Formula Weight | 225.29g/mol |
Odor | Odourless |
Packaging | Glass Bottle |
Color | Yellow |
pH | 5 to 7 (at 10.00 g/L 20°C) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'