Learn More
Dimethyl sulfoxide-d{6}, 99.5% (Isotopic), Thermo Scientific Chemicals
Catalog Number p-4408244
28.66 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-27-1
C2H6OS
84.17
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
75151
trideuterio(trideuteriomethylsulfinyl)methane
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
2206-27-1
C2H6OS
84.17
MFCD00002090
IAZDPXIOMUYVGZ-WFGJKAKNSA-N
dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
75151
trideuterio(trideuteriomethylsulfinyl)methane
[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.
Solubility
Miscible with water and with almost all organic solvents such as alcohols, esters, ketones, chlorinated solvents and aromatic hydrocarbons.
Notes
NMR solvent. Dimethyl Sulfoxide (DMSO) has a melting point of 20°C, freezing close to room temperature. Sometimes when it is delivered it will be in a solid state. To return the material to a liquid state, thaw it in a warm water bath. Care must be taken to prevent water contamination. It is stable, combustible and moisture sensitive. Incompatible with acid chlorides, strong acids, phosphorus halides, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Density 1.18 g/mL
Flash Point 88°C (190°F)
Refractive Index 1.476
Sensitivity Hygroscopic
Quantity 10 g
Melting Point 20°C
Boiling Point 190°C
Assay Percent Range 99.5%
Solubility Information Miscible with water and with almost all organic solvents such as alcohols,esters,ketones,chlorinated solvents and aromatic hydrocarbons.
Formula Weight 84.17
Chemical Name or Material Dimethyl Sulfoxide-d6