missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Dihydrexidine hydrochloride
GREENER_CHOICE

Product Code. 30001385
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 30001385

Brand: Tocris Bioscience™ 0884/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective D1-like agonist

A potent, full efficacy dopamine D1 agonist which shows no agonist activity at peripheral D2 receptors or adrenoceptors at doses which cause maximal stimulation of D1 sites. The compound appears to be fully bioavailable in brain and exhibits profound antiparkinsonism effects in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158704-02-0
Molecular Formula C17H18ClNO2

Specifications

Quantity 5 mg
Formula Weight 303.79
Purity >99%
Molecular Weight (g/mol) 303.786
Chemical Name or Material Dihydrexidine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.