Learn More
Binder™ Digital Pressure Display
Binder™ Digital Pressure Display had a measuring range from 1mbar to atmospheric pressure.
Brand:  Binder™ 8012-0806
Additional Details : Weight : 0.60000kg
 View more versions of this product
Product Code. 15621916
  • 1987.00€ / Each
please call +351214253350
Add to basket
Description
Description
  • Display accuracy of 1mbar
  • For use with either VD 53 or VD 115
Specifications
Specifications
VD 53
Digital pressure display
From 1mbar to atmospheric pressure
Videos