missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethylene glycol diacrylate, tech. 75%, stab., Thermo Scientific Chemicals
15.66 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.
Solubility
Slightly soluble in water (1-5g/100ml at 18°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.
Diethylene glycol diacrylate is used as reactive diluent and cross linking agent of radiation curing system, and also may be used as crosslinking resins, plastics and rubber modifier.
Solubility
Slightly soluble in water (1-5g/100ml at 18°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to light. Incompatible with strong oxidizing agents.
Specifications
Density | 1.118 |
Boiling Point | 94°C (0.2 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.462 |
Quantity | 25 g |
UN Number | UN2810 |
Beilstein | 1953711 |
Solubility Information | Slightly soluble in water (1-5g/100ml at 18°C). |
Formula Weight | 214.22 |
Percent Purity | 75% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'