missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dicyclohexyl phthalate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 63.0°C |
Color | White |
Density | 207g/mL |
Boiling Point | 200.0°C to 235.0°C (4.0 mmHg) |
Flash Point | 207°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: insoluble |
Show More |
Safety and Handling
Product Identifier
- Dicyclohexyl phthalate
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H361-Suspected of damaging fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- H317-May cause an allergic skin reaction.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'