missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dibenzyl ketoxime, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Dibenzyl ketoxime, is used as an intermediate for the preparation of cis-2-benzyl-3-phenylaziridine by reaction with lithium aluminum hydride. It is also used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | 122°C to 124°C |
Quantity | 5 g |
Beilstein | 1965977 |
Solubility Information | Insoluble in water. |
Formula Weight | 225.29 |
Percent Purity | ≥98% |
Chemical Name or Material | Dibenzyl ketoxime |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'