missing translation for 'onlineSavingsMsg'
Learn More

Diatomaceous Earth, Flux-Calcined, Honeywell Fluka™

Catalog Number p-7120369
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly Bottle
Poly Drum
This item is not returnable. View return policy
68855-54-9
CNa2O3
105.988
MFCD00132803
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]
This item is not returnable. View return policy
68855-54-9
CNa2O3
105.988
MFCD00132803
CDBYLPFSWZWCQE-UHFFFAOYSA-L
10340
CHEBI:29377
disodium;carbonate
C(=O)([O-])[O-].[Na+].[Na+]

flux-calcined, filter aid

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Packaging Poly Bottle
UN Number NONH for all modes of transport
Chemical Name or Material Diatomaceous Earth, Flux-Calcined
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'