Learn More
Diaion™ NSA100(Cl), ion exchange resin, highly cross-linked, strongly basic gel type I, 1.3 meq/ml on PS-DVB, Thermo Scientific™
Recovery of iodine and uranium
Brand: Thermo Scientific Alfa Aesar 046942.A1
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
300-1180μm Beads | |
Highly cross-linked, strongly basic gel type I, 1.3meq/mL on PS-DVB | |
300 to 1180μm | |
1 kg |
Diaion™ NSA100 (Cl), ion exchange resin | |
Odorless | |
White to Yellow |