Learn More
OHAUS™ Defender™ 3000 - I-D33 Bench Scale (EU)
Bench Scales for Basic Industrial Applications
625.00€ - 1130.00€
Specifications
Battery Life | 100 Hours with Rechargeable Battery |
---|---|
Battery Type | AC Adapter (Included);Rechargeable Battery (Included) |
Connection Type | Standard RS232 and optional USB Device or Ethernet |
Construction | Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet |
Display Type | 7 Segment LCD with Multicolor Backlight |
Product Code | Brand | Model | Readability | Capacity | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Readability | Capacity | Dimensions (L x W x H) | Price | Quantity & Availability | ||
17565825
|
OHAUS™
30685244 |
D33XW15B1R1 | 2 g | 15 kg | 489 x 305 x 569 mm |
625.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17575825
|
OHAUS™
30685245 |
D33XW30B1R1 | 5 g | 30 kg | 489 x 305 x 569 mm |
645.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17585825
|
OHAUS™
30685246 |
D33XW60B1R1 | 10 g | 60 kg | 489 x 305 x 569 mm |
645.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17595825
|
OHAUS™
30685247 |
D33XW60B1L2 | 10 g | 60 kg | 686 x 420 x 922 mm |
705.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17505835
|
OHAUS™
30685248 |
D33XW150B1L2 | 20 g | 150 kg | 686 x 420 x 922 mm |
705.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17515835
|
OHAUS™
30685249 |
D33XW150B1X2 | 20 g | 150 kg | 787 x 500 x 922 mm |
835.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17525835
|
OHAUS™
30685250 |
D33XW300B1X2 | 50 g | 300 kg | 787 x 500 x 922 mm |
835.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17535835
|
OHAUS™
30685251 |
D33XW300B1V3 | 50 g | 300 kg | 935 x 600 x 1222 mm |
1130.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17545835
|
OHAUS™
30685252 |
D33XW600B1V3 | 100 g | 600 kg | 935 x 600 x 1222 mm |
1130.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17555835
|
OHAUS™
30685253 |
D33XW15B1R5 | 2 g | 15 kg | 646 x 305 x 122 mm |
630.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17565835
|
OHAUS™
30685254 |
D33XW30B1R5 | 5 g | 30 kg | 646 x 305 x 122 mm |
630.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17575835
|
OHAUS™
30685255 |
D33XW60B1R5 | 10 g | 60 kg | 646 x 305 x 122 mm |
630.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17585835
|
OHAUS™
30685256 |
D33XW60B1L5 | 10 g | 60 kg | 842 x 420 x 134 mm |
755.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17595835
|
OHAUS™
30685257 |
D33XW150B1L5 | 20 g | 150 kg | 842 x 420 x 134 mm |
755.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17505845
|
OHAUS™
30685258 |
D33XW150B1X5 | 20 g | 150 kg | 941 x 500 x 150 mm |
920.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
17515845
|
OHAUS™
30685259 |
D33XW300B1X5 | 50 g | 300 kg | 941 x 500 x 150 mm |
920.00€
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
||
Description
- Results are clearly shown using the large, bright backlit LCD display.
- In Checkweighing mode display changes color (yellow/green/red) to alert when within the target weight range.
- This industrial scale features a 304 stainless steel platform, powder-coated steel tubular frame, and aluminum load cell.
- Indicator features a stainless steel housing and mounting bracket.
- Features software for Parts Counting, Checkweighing and Totalization.
- Includes RS232 with optional USB Device or Ethernet for printing or data connectivity.
Specifications
100 Hours with Rechargeable Battery | |
Standard RS232 and optional USB Device or Ethernet | |
7 Segment LCD with Multicolor Backlight | |
Weighing, Checkweighing, Counting, Totalization. |
AC Adapter (Included);Rechargeable Battery (Included) | |
Stainless steel indicator, 304-stainless steel platform, powder-coated steel tubular frame, aluminum load cell, non-slip adjustable rubber feet | |
-10°C to +40°C, 85%RH, non-condensing |