missing translation for 'onlineSavingsMsg'
Learn More

Decyldimethylchlorosilane, 97%, Thermo Scientific™

Catalog Number p-4228576
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
38051-57-9
C12H27ClSi
234.88
ZLZGHBNDPINFKG-UHFFFAOYSA-N
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
2758020
chloro-decyl-dimethylsilane
CCCCCCCCCC[Si](C)(C)Cl
This item is not returnable. View return policy
38051-57-9
C12H27ClSi
234.88
ZLZGHBNDPINFKG-UHFFFAOYSA-N
chloro decyl dimethylsilane, n-decyldimethylchlorosilane, decyldimethylchlorosilane, dimethyldecylchlorosilane, acmc-209ixd, 1-chlorodimethylsilyldecane, decyl-dimethyl-chlorosilane, 1-chlorodimethylsilyl decane, 10-chlorodecyl-dimethylsilicon, zlzghbndpinfkg-uhfffaoysa
2758020
chloro-decyl-dimethylsilane
CCCCCCCCCC[Si](C)(C)Cl
Color Colorless
Density 0.8660g/mL
Boiling Point 98°C (3.0 hPa)
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Quantity 5 mL
Specific Gravity 0.866
Formula Weight 234.88
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Decyldimethylchlorosilane
Show More Show Less
compliance-icons
Product Identifier
  • Decyldimethylchlorosilane, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'