Learn More
Afora™ Decanting Funnel with Cane
Separates immiscible, heterogenous liquids. Afora™ Decanting Funnel with Cane includes a plastic stopper and Rotaflo TF wrench.
Brand:  Afora™ 1098/TFE
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11921099
  • 93.53€ / Each
Estimated Shipment
29-04-2021
Add to basket
Description
Description
  • Pear-shaped body
  • Cone-shaped bottom
  • Glass with plastic stopcock
  • Includes Rotaflo TF wrench
Specifications
Specifications
122 mm
No
1000 mL
Glass