Learn More
Abnova™ DCN Recombinant Protein
Human DCN full-length ORF recombinant protein with GST-tag at N-terminal
335.00€ - 508.00€
Specifications
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
---|---|
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Molecular Weight (g/mol) | 65.23 |
pH Range | 8 |
Preparation Method | In vitro wheat germ expression system |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16113951
|
Abnova™
H00001634-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16123951
|
Abnova™
H00001634-P01.25UG |
25μg |
508.00€
25µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene is a small cellular or pericellular matrix proteoglycan that is closely related in structure to biglycan protein. The encoded protein and biglycan are thought to be the result of a gene duplication. This protein is a component of connective tissue, binds to type I collagen fibrils, and plays a role in matrix assembly. It contains one attached glycosaminoglycan chain. This protein is capable of suppressing the growth of various tumor cell lines. There are multiple alternatively spliced transcript variants known for this gene. This gene is a candidate gene for Marfan syndrome.
- Theoretical MW (kDa): 65.23
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MKATIILLLLAQVSWAGPFQQRGLFDFMLEDEASGIGPEVPDDRDFEPSLGPVCPFRCQCHLRVVQCSDLGLDKVPKDLPPDTTLLDLQNNKITEIKDGDFKNLKNLHALILVNNKISKVSPGAFTPLVKLERLYLSKNQLKELPEKMPKTLQELRAHENEITKVRKVTFNGLNQMIVIELGTNPLKSSGIENGAFQGMKKLSYIRIADTNITSIPQGLPPSLTELHLDGNKISRVDAASLKGLNNLAKLGLSFNSISAVDNGSLANTPHLRELHLDNNKLTRVPGGLAEHKYIQVVYLHNNNISVVGSSDFCPPGHNTKKASYSGVSLFSNPVQYWEIQPSTFRCVYVRSAIQLGNYK
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, Protein Array, ELISA, Western Blot | |
65.23 | |
In vitro wheat germ expression system | |
Wheat Germ (in vitro) | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Human | |
Solution |