missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-D-Lactose monohydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
Lactose is a disaccharide composed of glucose and galactose
Lactose is the major sugar found in dairy products such as milk and cheese
alpha-D-Lactose monohydrate is used as a carrier and stabilizer of products in the pharmaceutical and food industries
Applications
Lactose has been used in vitro to interact with carbohydrate recognition domains on a recombinant form of LGALS9, thus blocking downstream functions of the protein
It has been used as a control to study sugar dependency in Drosophila, which do not digest lactose
As a common component of pharmaceutical tablets, alpha-lactose has been used in laboratory settings to imitate drug administration
Specifications
Packaging | Plastic bottle |
Quantity | 250 g |
Formula Weight | 360.3 |
Chemical Name or Material | α-D-Lactose monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'