Learn More
D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D-Galactosamine is a pyranose amino sugar with hepatotoxic traits
• It acts as an RNA synthesis inhibitor in hepatocytes
• It promotes adherence of polymorphonuclear leukocytes to endothelial cells, increasing superoxide production
Applications
• D-Galactosamine can be used as a hepatotoxic in experimental in vitro models when in combination with LPS
• It is suitable for a range of biochemical laboratory applications
Specifications
Color | White |
Ignition Residue | 1% max. |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Quantity | 250 mg |
Specific Rotation Condition | +96° (20°C 589nm c=1,H2O,24 hr.) |
Specific Rotation | +93° to +99° (20°C, 589nm) (c=1, H2O), after 24 hrs. |
Formula Weight | 215.64 |
Percent Purity | ≥98.5% |
Show More |
RUO – Research Use Only