Learn More
D(+)-Galactosamine Hydrochloride, 99%, Thermo Scientific Chemicals
Catalog Number 11906201
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
1772-03-8
C6H13NO5·HCl
215.64
MFCD00135830
Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactose hydrochloride
1772-03-8
C6H13NO5·HCl
215.64
MFCD00135830
Chondrosamine hydrochloride, 2-Amino-2-deoxy-D-galactose hydrochloride
D(+)-Galactosamine hydrochloride ((2R,3R,4R,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal hydrochloride), CAS # 1772-03-8, is the salt of the amino sugar that acts as an RNA synthesis inhibitor in hepatocytes.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• D-Galactosamine is a pyranose amino sugar with hepatotoxic traits
• It acts as an RNA synthesis inhibitor in hepatocytes
• It promotes adherence of polymorphonuclear leukocytes to endothelial cells, increasing superoxide production
Applications
• D-Galactosamine can be used as a hepatotoxic in experimental in vitro models when in combination with LPS
• It is suitable for a range of biochemical laboratory applications
TRUSTED_SUSTAINABILITY
Melting Point 182°C to 185°C
Color White
Ignition Residue 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Merck Index 10,4205
Specific Rotation Condition +96° (20°C 589nm c=1,H2O,24 hr.)
Specific Rotation + 96
Solubility Information (10% in water) Clear colorless
Formula Weight 215.64
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Galactosamine hydrochloride