Learn More
D(+)-Fenchone, 97%, Thermo Scientific Chemicals
Catalog Number p-2691294
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
4695-62-9
C10H16O
152.24
MFCD00070689
LHXDLQBQYFFVNW-XCBNKYQSSA-N
+-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
1201521
CHEBI:165
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
4695-62-9
C10H16O
152.24
MFCD00070689
LHXDLQBQYFFVNW-XCBNKYQSSA-N
+-fenchone, 1s-1,3,3-trimethylbicyclo 2.2.1 heptan-2-one, 1s-1,3,3-trimethyl-norbornan-2-one
1201521
CHEBI:165
(1R,4S)-2,2,4-trimethylbicyclo[2.2.1]heptan-3-one
CC1(C2CCC(C2)(C1=O)C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 5°C to 7°C
Color Colorless to Yellow
Density 0.9500g/mL
Boiling Point 63°C to 65°C (13.0 mmHg)
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4610 to 1.464
Quantity 25 g
Beilstein 07, IV, 212
Merck Index 15, 4000
Specific Gravity 0.95
Specific Rotation Condition +59.00 (20.00°C neat)
Specific Rotation + 59
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in ethanol,ether,common organic solvents
Formula Weight 152.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material D(+)-Fenchone, 97%