missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus / Rhesus macaque BCMA / TNFRSF17 Protein, Fc Tag

Brand:  ACROBiosystems BCA-C5253-100UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15826027

  • 353.08€ / 100µg


Estimated Shipment: 26-10-2021
Add to basket

Description

Description

Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 32.1 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications

Specifications

XP_001106892.1
HEK293
Cynomolgus / Rhesus macaque
32.1 kDa
100 µg
Met 1 - Ala 53
TNFRSF17,CD269,BCM,BCMA
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>90% as determined by SDS-PAGE
Cynomolgus / Rhesus macaque BCMA, Fc Tag (BCA-C5253) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 53 (Accession # G7Q0I4-1). In the region Met 1 - Ala 53, the AA sequence of Cynomolgus and Rhesus macaque BCMA are homologus.
-20°C
BCMA
Fc Tag
Videos
SDS
Documents
Certificates