missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus PD-1 / PDCD1 Protein, Fc Tag

Brand:  ACROBiosystems PD1-C5254-100UG

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 15807457

  • 302.64€ / 100µg


Estimated Shipment: 29-10-2021
Add to basket

Description

Description

Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.6 kDa. The protein migrates as 53-60 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Specifications

Specifications

Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>90% as determined by SDS-PAGE
Cynomolgus PD-1, Fc Tag (PD1-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # B0LAJ3).
-20°C
PD-1
Fc Tag
HEK293
Cynomolgus
42.6 kDa
100 µg
Leu 25 - Gln 167
PDCD1,PD1,CD279,SLEB2
Unconjugated
50 mM Tris, 100 mM Glycine, pH7.5
Videos
SDS
Documents
Certificates