missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopropyl carbinol, 97+%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Specifications
CAS | 2516-33-8 |
Assay Percent Range | 97+% |
Molecular Formula | C4H8O |
MDL Number | MFCD00001309 |
Quantity | 25 g |
Synonym | cyclopropanemethanol, cyclopropyl carbinol, cyclopropylcarbinol, cyclopropylmethyl alcohol, hydroxymethyl cyclopropane, cyclopropanemethyl alcohol, cyclopropylcarbinyl alcohol, hydroxymethylcyclopropane, cyclopropyl-methanol, cyclopropane methanol |
InChI Key | GUDMZGLFZNLYEY-UHFFFAOYSA-N |
SMILES | OCC1CC1 |
IUPAC Name | cyclopropylmethanol |
Molecular Weight (g/mol) | 72.11 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'