Learn More
Cyclopropanecarboxaldehyde, 98%, Thermo Scientific Chemicals
Catalog Number p-2691281
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
1489-69-6
C4H6O
70.09
MFCD00012261
JMYVMOUINOAAPA-UHFFFAOYSA-N
cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
123114
cyclopropanecarbaldehyde
C1CC1C=O
1489-69-6
C4H6O
70.09
MFCD00012261
JMYVMOUINOAAPA-UHFFFAOYSA-N
cyclopropanecarboxaldehyde, formylcyclopropane, cyclopropylmethanal, cyclopropanecarboxyldehyde, cyclopropanal, cyclopropane aldehyde, cyclopropancarbaldehyd, cyclopropancarbaldehyde, cyclopropancarboxaldehyde, cyclopropanecarboaldehyde
123114
cyclopropanecarbaldehyde
C1CC1C=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.9380g/mL
Boiling Point 98.0°C to 101.0°C
Flash Point 7°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C3H5CHO
Refractive Index 1.4285 to 1.4305
Quantity 1 g
Specific Gravity 0.938
Solubility Information Solubility in water: insoluble.
Formula Weight 70.09
Percent Purity 98%
Chemical Name or Material Cyclopropanecarboxaldehyde, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopropanecarboxaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.