Learn More
Cyclohexyl methyl ketone, 95%
Product Code. 11480077
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11480077
Brand: Thermo Scientific Alfa Aesar B21000.06
Request Bulk or Custom Format
CAS: 823-76-7 | C8H14O | 126.199 g/mol
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It can be used to produce acetoxycyclohexane. It is also used as a pharmaceutical intermediate.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 823-76-7
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00040418
InChI Key RIFKADJTWUGDOV-UHFFFAOYSA-N
Synonym 1-cyclohexylethan-1-one, cyclohexyl methyl ketone, acetylcyclohexane, ethanone, 1-cyclohexyl, cyclohexylethanone, methyl cyclohexyl ketone, 1-acetylcyclohexane, cyclohexane, acetyl, acetophenone, hexahydro, ketone, cyclohexyl methyl
PubChem CID 13207
IUPAC Name 1-cyclohexylethanone
SMILES CC(=O)C1CCCCC1
Specifications
Melting Point -34°C
Density 0.917
Boiling Point 181°C to 183°C
Flash Point 66°C (151°F)
Assay Percent Range 0.95
Refractive Index 1.452
Quantity 5 g
Beilstein 507229
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 126.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Cyclohexyl methyl ketone
Show More Show Less
compliance-icons
Product Identifier
  • Cyclohexyl methyl ketone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Cyclohexylethan-1-one