Learn More
Cole-Parmer™ CPC™ Polysulfone Coupling
Features a smooth inner bore and leak-free seal protect against contamination
Brand: Cole-Parmer™ 31310-00
Additional Details : Weight : 0.22000kg
Description
- With their locking feature, these couplings guard against accidental disconnect and spills.
- To assemble, place the insert in the coupling body and twist to lock.
- An audible click ensures your connection.
- All couplings are made from USP Class VI-compliant polysulfone with platinum-cured silicone seals.
- Available in 3/4in. size, these fittings allow quick fill or empty of bioprocessing bags.
Specifications
Coupling Body | |
10/Pk. | |
Autoclave, Ethylene Oxide (ETO) or Gamma |
3/4 in. | |
Quick-Disconnect x Barb | |
Polysulfone |