Learn More
Helios Italquartz Covers with Long Handle, Translucent
Covers with long handle constructed of translucent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 64F00605
Additional Details : Weight : 0.20000kg
Description
- Translucent
- Include long handle
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Translucent | |
Quartz | |
Covers with Long Handle | |
CR 13 Crucible |