missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coronin 2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559458
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DIYPPTAGAQ PSLTAQEWLS GMNRDPILVS LRPGSELLRP HPLPAERPIF NSMAPASPRL LNQTEKLAAE DGW Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 66%.
Coronin 2A Polyclonal antibody specifically detects Coronin 2A in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
Coronin 2A | |
Polyclonal | |
Unconjugated | |
CORO2A | |
CLIPINB; CORO2A; coronin 2A; coronin, actin binding protein, 2A; coronin, actin-binding protein, 2A; coronin-2A; coronin-like protein B; IR10; WD protein IR10; WD repeat-containing protein 2; WDR2; WD-repeat protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92828 | |
CORO2A | |
Recombinant protein corresponding to Human Coronin 2A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only