man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Cell Culture Media, Supplements, and Reagents
Cell Culture Reagents, Antibiotics and Supplements
Antibiotics
Corning™ Gentamicin Sulfate
Print
Corning™ Gentamicin Sulfate
Packaged in plastic bottles
553.85€
See promotional offers below
Specifications
Product Type
Antibiotic
Sterility
Sterile
Format
Plastic Bottle
View More Specs
Products
1
Documents
Product Certifications
Products
1
Product Code
Brand
Quantity
Form
Price
Quantity & Availability
Product Code
Brand
Quantity
Form
Price
Quantity & Availability
15353671
View Documents
Promotion Details
missing translation for 'viewProductCertificates'
Corning™
30-005-CR
10 x 10 mL
Liquid
553.85€
10mL
View alternative products
Save Up to
»
Estimated Shipment: 20-06-2024
Log in
Specifications
Product Type
Antibiotic
Sterility
Sterile
Format
Plastic Bottle
Documents
Product Certifications
Back to Top