Learn More
CoraLite™ Plus 488 Rat IgG2a Isotype Control (2A3), Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL488-65209-100UG
This item is not returnable.
View return policy
Description
The IgG2a Isotype Control antibody from Proteintech is a rat monoclonal antibody generated with n/a of rat IgG2a Isotype Control. This antibody recognizes antigen. The IgG2a Isotype Control antibody has been validated for the following applications: FC analysis. This antibody is coralite™ plus 488.Specifications
| IgG2a Isotype Control | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| 100 μg | |
| Primary | |
| Store at 2-8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Flow Cytometry | |
| 2A3 | |
| CoraLite Plus 488 | |
| Affinity purification | |
| RUO | |
| Not Applicable | |
| Flow Antibody | |
| IgG2a κ |