Learn More
Copper(II) Sulfate Anhydrous, Extra Pure, SLR, Fisher Chemical™
Catalog Number p-7072950
Click to view available options
Quantity:
250 g
1 kg
7758-98-7
CuO4S
159.60
10981
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper(2+) sulfate
[Cu++].[O-]S([O-])(=O)=O
7758-98-7
CuO4S
159.60
10981
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper(2+) sulfate
[Cu++].[O-]S([O-])(=O)=O
Melting Point 200°C
Color Gray
pH 3.5 to 4.5
Physical Form Powder/Solid
Quantity 250 g
Formula Weight 159.6g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.