missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) pyrithione, Thermo Scientific Chemicals

Catalog Number p-7077661
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14915-37-8
C10H8CuN2O2S2
315.86
This item is not returnable. View return policy
14915-37-8
C10H8CuN2O2S2
315.86

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 315.86
Percent Purity ≥97.5% (HPLC)
Infrared Spectrum Authentic
Chemical Name or Material Copper(II) pyrithione

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'