missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Test Solution Potassium Chloride, SI Analytics™

Catalog Number 11904789
Click to view available options
Quantity:
20 mL
250 mL
500 mL
Packaging:
3 x 6 Ampules
Polyethylene Bottle
Concentration:
0.01/0.1/1mol/L (1.41/12.9/112mS/cm)
0.01mol/L (1.41 mS/cm)
0.1mol/L (12.9mS/cm)
5mS/cm
This item is not returnable. View return policy
This item is not returnable. View return policy

SI Analytics™ Conductivity Test Solution Potassium Chloride allows for the determination of a substances ability to conduct electricity.

  • Mixture consisting of water and potassium chloride
  • Store at room temperature
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Product does not present an explosion hazard
  • Do not allow undiluted product or large quantities of it to reach ground water, water course or sewage system
  • Small amounts may be diluted with plenty of water and washed away
Physical Form Fluid
Concentration 0.1mol/L (12.9mS/cm)
Odor Odorless
Packaging 3 x 6 Ampules
Density 1g/cm3
pH 7
Melting Point -0.3°C
Boiling Point 100°C
Color Colorless
Quantity 20 mL
Solubility Miscible with water
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'