Learn More
Colorimetric Standard Solution, Yellow, Chem Lab™
Catalog Number 11997423
Click to view available options
Quantity:
100 mL
Quantity 100 mL
Color Yellow
Physical Form Liquid
compliance-icons
Product Identifier
  • Colour stock standard solution Y (yellow)
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iron chloride, cobalt chloride, hydrochloric acid