Learn More
Sampling Systems™ Cohesive Sampler
Constructed of robust stainless steel and able to take a sample from a known point within the product. Sampling Systems™ Cohesive Sampler features a scraper that is fully welded to the outside of the pocket.
775.00€ - 790.00€
Specifications
Product Type | Sampler |
---|---|
Material | Stainless Steel (316L) |
Length (Metric) | 1000 mm |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15809311
|
Sampling Systems™
2015A-05 |
5 mL |
775.00€
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
15819311
|
Sampling Systems™
2015A-10 |
10 mL |
780.00€
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
15829311
|
Sampling Systems™
2015A-25 |
25 mL |
790.00€
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||||
Description
- Manufactured in 316L Stainless Steel
- Single point type
- Used to push the powder into the sampler by rotating it
Specifications
Sampler | |
Stainless Steel (316L) | |
1000 mm |