missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTNAP5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567034
Additional Details : Weight : 0.01000kg
Description
May play a role in the correct development and proper functioning of the peripheral and central nervous system and be involved in cell adhesion and intercellular communication.Specifications
| CNTNAP5 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WYK1 | |
| CASPR5 | |
| Rabbit | |
| IgG | |
| 100 μL | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Primary | |
| 129684 |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| CNTNAP5 | |
| Liquid | |
| CNTNAP5 | |
| Recombit Human CNTNAP5 | |
| Affinity Chromatography | |
| RUO | |
| Antibody | |
| Polyclonal | |
| Human |
For Research Use Only