missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOX Antibody [Alexa Fluor™ 594], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-46570AF594
This item is not returnable.
View return policy
Description
CNOX Polyclonal antibody specifically detects CNOX in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| CNOX | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55068 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| Alexa Fluor 594 | |
| candidate growth-related and time keeping constitutive hydroquinone (NADH)oxidase, Candidate growth-related and time keeping constitutive hydroquinone [NADH]oxidase, cCNOXConstitutive Ecto-NOX, Cell proliferation-inducing gene 38 protein, cNOX, CNOXbA64J21.1, ecto-NOX disulfide-thiol exchanger 1, FLJ10094, PIG38, proliferation-inducing protein 38 | |
| ecto-NOX disulfide-thiol exchanger 1. (Uniprot# Q8TC92) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction