missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Cisapride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
5-HT4 receptor agonist and gastrokinetic agent. Stimulates intestinal acetylcholine release, possibly via 5-HT4 receptor-dependent and -independent mechanisms, leading to increased intestinal motility.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 465.95 |
| Purity | >99% |
| Molecular Weight (g/mol) | 465.95 |
| Chemical Name or Material | Cisapride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction