Learn More
CHEMetrics™ Chemets™ Water Testing Kits
Provide results in two minutes or less
Brand: CHEMetrics™ K6904
Additional Details : Weight : 0.80000kg
Includes: Complete test kits contain 30 ampules of reagent, comparator(s), snap cup (holds water sample), instructions, and MSDS sheets. All components are housed in a sturdy, plastic case
Description
Provide results in two minutes or less
Water testing kits for Chlorine (DPD), Copper, Dissolved Oxygen, Hydrogen Peroxide, Phenols, Phosphate, Silica, Sulfide and more. Designed for maximum simplicity and accuracy, Chemets 7mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside.
- To perform a test, simply immerse Chemets ampule in the sample, snap off tip, and the correct sample volume is quickly drawn inside the ampule
- In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
Comparators have two-year shelf life.
Order Info
Refill packs of 30 ampules are also available.
Specifications
Nitrate | |
Chemets™ Water Testing Kit | |
Water testing kits for chlorine (DPD), copper, dissolved oxygen, hydrogen peroxide, phenols, phosphate, silica, sulfide and more | |
30.0 | |
Cadmium Reduction |
Nitrate | |
0 to 3ppm as N | |
Plastic case, 30 ampoules of reagent, refill, comparators, 25mL sample cup, instructions, reaction tube with lid | |
15 min. |
Safety and Handling
- CHEMets nitrate 0-3PPM
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Specific target organ toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 1B
Hazard Statement
- H330-Fatal if inhaled.
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H372-Causes damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P101-If medical advice is needed, have product container or label at hand.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P102-Keep out of reach of children.
- P271-Use only outdoors or in a well-ventilated area.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P270-Do not eat, drink or smoke when using this product.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P273-Avoid release to the environment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P310-Immediately call a POISON CENTER/doctor/.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P405-Store locked up.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to authourised chemicallandfill or if organic to high temperature incineration.
Supplemental information
- MIXTURE LIST-Contains : tartaric acid, sodium sulphate, sodium tartrate, cyclohexanediaminetetraacetic acid, trisodiium salt