Learn More
Honeywell™ Check and Go DeepTril 3 Gloves
Click to view available options
Size:
7
8
9
10
11
Description
- Black HPPE knitted glove
- Fully dipped black foam nitrile coating
- Elasticated knitted wrist
- The ergonomics seamless knit construction conforms better to the natural shape of the hand for greater strength and durability, superior confort
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The foam coating optimise the glove's breathability
- The nitrile foam and PVC dots on palm improves the grip
- The knitted and elasticated wrist gives the glove good support
- The nitrile coating provides outstanding abrasion resistance and a good protection in light oily environement
- HPPE provides very good resistance to cut
- The black color is suitable for work in a soiled atmosphere
Specifications
Certifications/Compliance | EN 420-2003 Basic standards, EN 388-2003 Mechanical hazards |
Description | 13 gauge |
Color | Black |
Material | HPPE, Nitrile |
Size | 7 |
Length (Metric) | 25 cm |