Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Marque: Thermo Scientific™ FCR400TSXLE
Informations supplémentaires : Weight : 13.00000kg
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6″ (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorderPrestations de service
Warranty varies by country, please contact us for details
Spécification
Inkless Chart Recorder, Field Installed | |
Chart Recorder | |
TSX400 ultra-low freezers | |
Recorder |