missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cesium Hydroxide Monohydrate, Honeywell Fluka™
Click to view available options
Quantity:
10 g
50 g
250 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 250 g |
UN Number | 2682 |
IUPAC Name | caesium(1+) hydrate hydroxide |
Formula Weight | 167.93g/mol |
Percent Purity | ≥95% (T) |
Density | 3.68g/mL at 25°C |
Chemical Name or Material | Cesium Hydroxide Monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'