Learn More
Honeywell Fluka 100GR Celite 545 AW filter aid, acid washed, treated with sodium carbonate, flux calcined
Brand: Honeywell Fluka 22141-100G
100GR Celite 545 AW filter aid, acid washed, treated with sodium carbonate, flux calcined
Additional Details : CAS : 68855-54-9 Weight : 0.10000kg