missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cefmenoxime Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Cefmenoxime HCl |
| CAS | 75738-58-8 |
| Molecular Formula | 2 C16 H17 N9 O5 S3 . Cl H |
| Quantity | 100 mg |
| Synonym | 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2Z)-(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, hydrochloride (2:1), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2-amino-4-thiazolyl)(methoxyimino)acetyl]amino]-3-[[(1-methyl-1H-tetrazol-5-yl)thio]methyl]-8-oxo-, hydrochloride (2:1), [6R-[6α,7β(Z)]]-, Abbott 50192, Bestcall, Bestron, Cefmax, Cefmenoxime hemihydrochloride, Cefmenoxime hydrochloride, Cemix, Tacef |
| InChI Formula | InChI=1S/2C16H17N9O5S3.ClH/c2*1-24-16(20-22-23-24)33-4-6-3-31-13-9(12(27)25(13)10(6)14(28)29)19-11(26)8(21-30-2)7-5-32-15(17)18-7;/h2*5,9,13H,3-4H2,1-2H3,(H2,17,18)(H,19,26)(H,28,29);1H/b2*21-8-;/t2*9-,13-;/m11./s1 |
| SMILES | Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)O)CSc3nnnn3C)\c4csc(N)n4.CO\N=C(/C(=O)N[C@H]5[C@H]6SCC(=C(N6C5=O)C(=O)O)CSc7nnnn7C)\c8csc(N)n8 |
| IUPAC Name | (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrochloride |
| Molecular Weight (g/mol) | 1059.58 |
| Formula Weight | 1058.0796 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?