Learn More
CD68 Rat anti-Mouse, PE, Clone: FA/11 (also known as FA-11, FA11), BD Pharmingen™
Rat Monoclonal Antibody
Brand: BD Biosciences 566386
Additional Details : Weight : 0.00900kg
Description
CD68 Monoclonal antibody specifically detects CD68 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD68 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
FA/11 (also known as FA-11, FA11) | |
PE | |
0.1 mg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |