missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse anti-Human, PE, Clone: UCHT2, BD
Mouse Monoclonal Antibody
189.00€ - 415.00€
Specifications
Antigen | CD5 |
---|---|
Clone | UCHT2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15821449
|
BD Biosciences
561897 |
25 Tests |
189.00€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
15878088
|
BD Biosciences
555353 |
100 Tests |
415.00€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
Description
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
Specifications
CD5 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
CD5 antigen (p56-62); T1; Tp67; LEU1; Lymphocyte antigen T1/Leu-1 |
UCHT2 | |
Monoclonal | |
Mouse | |
Human | |
Human T cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.