missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CD44 Pre-design Chimera RNAi
Homo sapiens CD44 antigen (homing function and Indian blood group system) (CD44), transcript variant 8, mRNA.
POA
Specifications
Content And Storage | Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
---|---|
For Use With (Application) | RNAi Knockdown |
Description
- Gene name: CD44
- Gene Alias: CDW44,CSPG8,ECMR-III,HCELL,IN,LHR,MC56,MDU2,MDU3,MGC10468,MIC4,MUTCH-I,Pgp1
- Entrez GeneID: 960
- Gene Description: CD44 molecule
Specifications
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. | |
RNAi Knockdown |
For research use only