missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4 Rat anti-Mouse, BB700, Clone: RM4-5, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566408
Additional Details : Weight : 0.01000kg
Description
CD4 Monoclonal antibody specifically detects CD4 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD4 | |
Monoclonal | |
0.2 mg/mL | |
Rat | |
RUO | |
Mouse | |
Purified |
Flow Cytometry | |
RM4-5 | |
Brilliant Blue 700 (BB700) | |
25 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG2a κ |