missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse anti-Human, BB700, Clone: HIT2, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566445
Additional Details : Weight : 0.00840kg
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry.Specifications
CD38 | |
Monoclonal | |
Brilliant Blue 700 (BB700) | |
100 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
HIT2 | |
Mouse | |
RUO | |
CD38 | |
Refrigerate (2 to 8°C) |