Learn More
CD244.2 Mouse anti-Mouse, FITC, Clone: 2B4, BD
Mouse Monoclonal Antibody
POA
Specifications
Antigen | CD244.2 |
---|---|
Clone | 2B4 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD244 is a type I transmembrane glycoprotein that is also known as Natural killer cell receptor 2B4 (NKR2B4), or Signaling lymphocytic activation molecule 4 (SLAMF4). CD244 is a member of the CD2 subset of the immunoglobulin superfamily (CD2 IgSF). It is expressed on all natural killer (NK) cells, IL-2-activated NK (LAK) cells, committed progenitors of NK cells, and a subset of T lymphocytes which mediate non-MHC-restricted cytoxicity, including dendritic epidermal T cells. The 2B4 monoclonal antibody specifically binds to CD244.2, the 2B4 alloantigen which is expressed on C57BL/6 and C58/J mice, but not in most strains tested (A/J, AKR, BALB/c, CBA/J, CBA/N, C3H/He, C57BR, DBA/1, DBA/2, NZB, SJL/J, 129). At least two isoforms of CD244.2 protein, products of alternative splicing of hnRNA, are expressed on IL-2-activated C57BL/6 NK cells. They differ only in their cytoplasmic domains; with 2B4L (150-aa cytoplasmic tail) having inhibitory activity and 2B4S (93-aa tail) being stimulatory. The extracellular domain of CD244 is a ligand for another CD2 IgSF member, CD48. 2B4 antibody activates lytic and secretory functions of IL-2-cultured NK cells and CD244.2+ T cells.
Specifications
CD244.2 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
rIL-2-propagated NK1.1+ cells from C57BL/6 mice | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
2B4 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified | |
CD242.2; 2B4 B6 Alloantigen; Ly90; NAIL; Nmrk; SLAMF4 |
For Research Use Only.