missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD20 Mouse anti-Human, PE-Cy5, Clone: 2H7, BD
Mouse Monoclonal Antibody
249.00€ - 471.00€
Specifications
Antigen | CD20 |
---|---|
Clone | 2H7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-Cyanine5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15861309
|
BD Biosciences
561761 |
25 Tests |
249.00€
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
15808318
|
BD Biosciences
555624 |
100 Tests |
471.00€
Each |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.
Specifications
CD20 | |
Flow Cytometry | |
PE-Cyanine5 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2b κ | |
Affinity Purified |
2H7 | |
Monoclonal | |
Mouse | |
Human | |
Human 6.16c1.3 B cell line | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.